Learn More
Medchemexpress LLC WYE-687 | 1062161-90-3 | C28H32N8O3 | 5 MG
SDP
Catalog No. 5000372345
Encompass_Preferred
WYE-687 is an ATP-competitive mTOR inhibitor with an IC50 of 7 nM. It concurrently inhibits the activation of mTORC1 and mTORC2. WYE-687 also inhibits PI3Kα and PI3Kγ with IC50s of 81 nM and 3.11 μM, respectively.
  • ATP-competitive mTOR inhibitor
  • Inhibits activation of mTORC1 and mTORC2
  • Inhibits PI3Kα and PI3Kγ
  • Potently inhibits HL-60 cell survival
  • Cytotoxic to U937, THP-1 and AML-193 AML cell lines
  • Significantly inhibits U937 xenograft tumor growth in SCID mice
Catalog No. 50-003-72345 Supplier Medchemexpress LLC Supplier No. HY152715MG
Add to Cart
Add to Cart
WYE-687 is an ATP-competitive mTOR inhibitor with an IC50 of 7 nM. It concurrently inhibits the activation of mTORC1 and mTORC2. WYE-687 also inhibits PI3Kα and PI3Kγ with IC50s of 81 nM and 3.11 μM, respectively.
  • ATP-competitive mTOR inhibitor
  • Inhibits activation of mTORC1 and mTORC2
  • Inhibits PI3Kα and PI3Kγ
  • Potently inhibits HL-60 cell survival
  • Cytotoxic to U937, THP-1 and AML-193 AML cell lines
  • Significantly inhibits U937 xenograft tumor growth in SCID mice