Learn More
Med Vet International Wye-Connector Rigid, J0249X
Catalog No. 5000582997
Encompass_Preferred
Two legs fit standard 22mm anesthetic hoses, and third legs fits 15mm endotracheal tube connectors.
Catalog No. 50-005-82997 Supplier Med Vet International Supplier No. JOR249X
Two legs fit standard 22mm anesthetic hoses, and third legs fits 15mm endotracheal tube connectors.