Learn More
Med Vet International Wye-Connector, Swivel, J0249Y
Catalog No. 5000582224
Encompass_Preferred
Two legs fit standard 22mm anesthetic hoses, neutral-colored nylon; third leg fits 15mm endotracheal tube connector.
Catalog No. 50-005-82224 Supplier Med Vet International Supplier No. JOR249Y
Two legs fit standard 22mm anesthetic hoses, neutral-colored nylon; third leg fits 15mm endotracheal tube connector.