Learn More
Kimberly-Clark Professional™ WypAll™ Disinfecting Wipes
Shop All Kimberly-Clark Professional Products

Description
- WypAll™ Disinfecting Wipes offer a clean you can trust with chemicals you can pronounce - at a price you can afford.
- Activated with biodegradable hydrogen peroxide, the clean, fresh-scented WypAll™ formula packs a punch by killing pathogens such as Norovirus and the COVID-19 virus while being approved for use in commercial, industrial, and healthcare settings.
- It is also designed to meet the lowest EPA-defined human toxicity levels for disinfecting wipes, which means they can be labeled without the Hazards to Humans or Wash Hands After Use warnings typically noted on the vast majority of disinfecting wipes canisters.
- Configured for businesses, WypAll™ Disinfecting Wipes come in canisters with a high sheet count.
- WypAll™ Disinfecting Wipes deliver a reasonable cost per wipe with efficient storage, making it easier to handle for your people, the planet, and your pocketbook.
Specifications
Specifications
| Product Type | Disinfectant Wipe |
| Length (English) | 15.875 in. |
| Length (Metric) | 15.875 in. |
| Width (English) | 5.438 in. |
| Width (Metric) | 5.438 in. |
| Material | Non-woven Polypropylene |
| Autoclavable | Not Autoclavable |
| Disposable | Single-use |
| Dimensions (L x W) | 15.875 x 5.438 in. |
| For Use With (Application) | Surface disinfectant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.