Learn More
Poltex Wypall Dispenser X60
SDP
Catalog No. 22022530
Change view
Click to view available options
Mounting:
Magnetic Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-022-530 Magnetic Mount
22-022-529 VHB Tape
22-022-528 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-022-530 Supplier Poltex Supplier No. WYPL34790M
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Dispenser holds and dispenses Wypall x60, #34790 wipes. Durable PETG construction.
  • Holds and dispenses Wypall x60, #34790 wipes
  • Durable PETG construction
  • Available as magnet mount (powerful magnets included), wall mount (screws/anchors included), and tape mount (VHB tape included)
Specifications
Product Type Wipe Dispenser
Color Clear
Length (Metric) 127 mm
Width (Metric) 248.92 mm
Height (Metric) 248.92 mm
Holds Wipes, Labwipes
Material PETG
Mounting Magnetic Mount
No. of Compartments 1
Product Style Side Loading
Sterility Non-sterile