Learn More
Grainger WYPALL, Dry Wipe 9INX16.75IN 126/EA
Catalog No. NC1865736
Encompass_Preferred
The Wypall Dry Wip is a versatile, disposable cleaning cloth. Made from hydroknit, it's low-linting with a textured surface for effective cleaning. At 12" x 12", these durable cloths come in 150-packs, suitable for heavy-duty or delicate surfaces. They effectively trap dirt and can be used wet or dry on glass, metal, plastic, and more, ideal for various industrial settings.
This item is not returnable. View return policy
Catalog No. NC1865736 Supplier Grainger Supplier No. 2VHP9
The Wypall Dry Wip is a versatile, disposable cleaning cloth. Made from hydroknit, it's low-linting with a textured surface for effective cleaning. At 12" x 12", these durable cloths come in 150-packs, suitable for heavy-duty or delicate surfaces. They effectively trap dirt and can be used wet or dry on glass, metal, plastic, and more, ideal for various industrial settings.