Learn More
Medchemexpress LLC HY-12001 5mg Medchemexpress, WZ-3146 CAS:1214265-56-1 Purity:>98%
SDP
Catalog No. 501932545
Encompass_Preferred
Medchemexpress, HY-12001 5mg WZ-3146 CAS:1214265-56-1 WZ3146 is a mutant selective EGFR inhibitor with IC 50 s of 2, 2, 5, 14 and 66 nM for EGFR L858R , EGFR L858R/T790M , EGFR E746_A750 , EGFR E746_A750/T790M and EGFR, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2545 Supplier Medchemexpress LLC Supplier No. HY120015MG
Medchemexpress, HY-12001 5mg WZ-3146 CAS:1214265-56-1 WZ3146 is a mutant selective EGFR inhibitor with IC 50 s of 2, 2, 5, 14 and 66 nM for EGFR L858R , EGFR L858R/T790M , EGFR E746_A750 , EGFR E746_A750/T790M and EGFR, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.