Learn More
Apexbio Technology LLC WZ4002, 50mg. Cas: 1213269-23-8 MFCD: MFCD16621244. Mutant-selective EGFR inhibitor(L858R,T790M), irreversible and potent
SDP
Supplier:  Apexbio Technology LLC A138950
Encompass_Preferred
WZ4002 is a novel mutant-selective epidermal growth factor receptor (EGFR) kinase inhibitor that potently and irreversibly suppresses the ATP-dependent auto-phosphorylation of both wild-type EGFR and EGFR mutants. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4697