Learn More
Medchemexpress LLC HY-12026 50mg Medchemexpress, WZ4002 CAS:1213269-23-8 Purity:>98%
SDP
Catalog No. 501969596
Encompass_Preferred
Medchemexpress, HY-12026 50mg WZ4002 CAS:1213269-23-8 WZ4002 is a mutant selective EGFR inhibitor with IC50s of 2, 8, 3 and 2 nM for EGFRL858R, EGFRL858R/T790M, EGFRE746_A750 and EGFRE746_A750/T790M, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9596 Supplier Medchemexpress LLC Supplier No. HY1202650MG
Medchemexpress, HY-12026 50mg WZ4002 CAS:1213269-23-8 WZ4002 is a mutant selective EGFR inhibitor with IC50s of 2, 8, 3 and 2 nM for EGFRL858R, EGFRL858R/T790M, EGFRE746_A750 and EGFRE746_A750/T790M, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.