Learn More
Medchemexpress LLC HY-12026 100mg , WZ4002 CAS:1213269-23-8 Purity:98%
SDP
Catalog No. 501493243
Encompass_Preferred
Medchemexpress, HY-12026 100mg WZ4002 CAS:1213269-23-8 Formula:C25H27ClN6O3 IC50: 2 nM (EGFR L858R ), 8 nM (EGFR L858R/T790M ), 3 nM (EGFR E746_A750 ), 2 nM (EGFR E746_A750/T790M ) Purity:98% WZ4002 is a mutant selective EGFR inhibitor with IC 50 s of 2, 8, 3 and 2 nM for EGFR L858R , EGFR L858R/T790M , EGFR E746_A750 and EGFR E746_A750/T790M , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3243 Supplier Medchemexpress LLC Supplier No. HY12026100MG
Medchemexpress, HY-12026 100mg WZ4002 CAS:1213269-23-8 Formula:C25H27ClN6O3 IC50: 2 nM (EGFR L858R ), 8 nM (EGFR L858R/T790M ), 3 nM (EGFR E746_A750 ), 2 nM (EGFR E746_A750/T790M ) Purity:98% WZ4002 is a mutant selective EGFR inhibitor with IC 50 s of 2, 8, 3 and 2 nM for EGFR L858R , EGFR L858R/T790M , EGFR E746_A750 and EGFR E746_A750/T790M , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.