Learn More
Selleck Chemical LLC WZ4002 10mg 1213269-23-8
SDP
Catalog No. 508289
Encompass_Preferred
WZ4002 is a novel, mutant-selective EGFR inhibitor for EGFR(L858R)/(T790M) with IC50 of 2 nM/8 nM in BaF3 cell line; does not inhibit ERBB2 phosphorylation (T798I). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-828-9 Supplier Selleck Chemical LLC Supplier No. S117310MG
WZ4002 is a novel, mutant-selective EGFR inhibitor for EGFR(L858R)/(T790M) with IC50 of 2 nM/8 nM in BaF3 cell line; does not inhibit ERBB2 phosphorylation (T798I). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.