Learn More
Apexbio Technology LLC WZ4003 1214265-58-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061517
Encompass_Preferred
WZ4003 (CAS 1214265-58-3) is a selective inhibitor targeting NUAK1 and NUAK2 kinases members of the AMP-activated protein kinase (AMPK) family activated by tumor suppressor kinase LKB1 WZ4003 inhibits NUAK1 and NUAK2 with IC50 values of 20 nM and 100 nM respectively In cellular assays treatment with WZ4003 reduces NUAK1-mediated phosphorylation of MYPT1 at Ser445 It suppresses cell proliferation migration of mouse embryonic fibroblasts and invasion of U2OS cells Thus WZ4003 serves as a valuable tool for investigating NUAK kinase functions in cell proliferation mobility and signaling mechanisms relevant to oncology research
Catalog No. 50-000-61517 Supplier Apexbio Technology LLC Supplier No. B13745.1
WZ4003 (CAS 1214265-58-3) is a selective inhibitor targeting NUAK1 and NUAK2 kinases members of the AMP-activated protein kinase (AMPK) family activated by tumor suppressor kinase LKB1 WZ4003 inhibits NUAK1 and NUAK2 with IC50 values of 20 nM and 100 nM respectively In cellular assays treatment with WZ4003 reduces NUAK1-mediated phosphorylation of MYPT1 at Ser445 It suppresses cell proliferation migration of mouse embryonic fibroblasts and invasion of U2OS cells Thus WZ4003 serves as a valuable tool for investigating NUAK kinase functions in cell proliferation mobility and signaling mechanisms relevant to oncology research