Learn More
AdipoGen WZB117
SDP
Catalog No. 502562211
Encompass_Preferred
Chemical. CAS 1223397-11-2. Formula C20H13FO6. MW 368.3. . GLUT inhibitor GLUT4 > GLUT1 = GLUT3. Fast-acting irreversible glucose transporter 1 GLUT1 inhibitor. Binds directly to GLUT1 involving three hydrogen bonds, one each with Asn34, Arg126, and Trp412. Blocks glucose transport in diverse cancer cells, reducing extracellular lactate and intracellular ATP levels and consequently decreases glycolysis. Anticancer agent in vitro and in vivo. Inhibits cancer cell proliferation, resulting in cell cycle arrest, senescence and necrosis or apoptosis. Inhibits the self-renewal and tumor-initiating capacity of cancer stem cells, blocking tumor initiation after implantation of cancer stem cells in mice.
Catalog No. 50-256-2211 Supplier AdipoGen Supplier No. AGCR13694M025
Chemical. CAS 1223397-11-2. Formula C20H13FO6. MW 368.3. . GLUT inhibitor GLUT4 > GLUT1 = GLUT3. Fast-acting irreversible glucose transporter 1 GLUT1 inhibitor. Binds directly to GLUT1 involving three hydrogen bonds, one each with Asn34, Arg126, and Trp412. Blocks glucose transport in diverse cancer cells, reducing extracellular lactate and intracellular ATP levels and consequently decreases glycolysis. Anticancer agent in vitro and in vivo. Inhibits cancer cell proliferation, resulting in cell cycle arrest, senescence and necrosis or apoptosis. Inhibits the self-renewal and tumor-initiating capacity of cancer stem cells, blocking tumor initiation after implantation of cancer stem cells in mice.