Learn More
Medchemexpress LLC X-376 | 1365267-27-1 | 98.1% | 547.41 g·mol⁻¹ | C25H25Cl2FN6O3 | 10 MG
SDP
Catalog No. 5000289182
Encompass_Preferred
X-376 is a small-molecule tyrosine kinase inhibitor that potently targets anaplastic lymphoma kinase (ALK) and shows inhibitory activity against MET. It is supplied as a powder for preclinical research use and is suitable for cellular and in vivo studies of ALK-driven tumors.
  • Potent ALK inhibition (IC50 0.61 nM).
  • MET inhibitory activity (IC50 0.69 nM).
  • Suitable for preclinical cellular and in vivo research.
  • Supplied as a powder with recommended cold storage for stability.
  • Molecular weight 547.41 g·mol⁻¹; formula C25H25Cl2FN6O3.
Catalog No. 50-002-89182 Supplier Medchemexpress LLC Supplier No. HY1659010MG
X-376 is a small-molecule tyrosine kinase inhibitor that potently targets anaplastic lymphoma kinase (ALK) and shows inhibitory activity against MET. It is supplied as a powder for preclinical research use and is suitable for cellular and in vivo studies of ALK-driven tumors.
  • Potent ALK inhibition (IC50 0.61 nM).
  • MET inhibitory activity (IC50 0.69 nM).
  • Suitable for preclinical cellular and in vivo research.
  • Supplied as a powder with recommended cold storage for stability.
  • Molecular weight 547.41 g·mol⁻¹; formula C25H25Cl2FN6O3.