Learn More
Medchemexpress LLC X-376 | 1365267-27-1 | 98.10% | 547.41 | 25 MG
SDP
Catalog No. 5000394996
Encompass_Preferred
X-376 is a potent and highly specific ALK tyrosine kinase inhibitor (TKI) with an IC50 of 0.61 nM. It is also a less potent inhibitor of MET (IC50=0.69 nM) and exhibits potent anti-tumor activity.
  • Effective in inhibiting the growth of various cancer cell lines, including H3122 lung cancer cells (IC50: 77 nM), H2228 lung cancer cells (IC50: 57 nM), and SUDHL-1 lymphoma cells (IC50: 32 nM).
  • Also inhibits SY5Y neuroblastoma cells, MKN-45 gastric carcinoma cells, HepG2 cells, and PC-9 lung cancer cell lines.
  • Demonstrates substantial bioavailability and moderate half-lives in vivo.
  • Significantly delays tumor growth in H3122 xenografts at a dose of 50 mg/kg bid, appearing well-tolerated without affecting mouse weight or causing compound-related toxicity.
Catalog No. 50-003-94996 Supplier Medchemexpress LLC Supplier No. 25MGHY16590
X-376 is a potent and highly specific ALK tyrosine kinase inhibitor (TKI) with an IC50 of 0.61 nM. It is also a less potent inhibitor of MET (IC50=0.69 nM) and exhibits potent anti-tumor activity.
  • Effective in inhibiting the growth of various cancer cell lines, including H3122 lung cancer cells (IC50: 77 nM), H2228 lung cancer cells (IC50: 57 nM), and SUDHL-1 lymphoma cells (IC50: 32 nM).
  • Also inhibits SY5Y neuroblastoma cells, MKN-45 gastric carcinoma cells, HepG2 cells, and PC-9 lung cancer cell lines.
  • Demonstrates substantial bioavailability and moderate half-lives in vivo.
  • Significantly delays tumor growth in H3122 xenografts at a dose of 50 mg/kg bid, appearing well-tolerated without affecting mouse weight or causing compound-related toxicity.