Learn More
Torpac Inc X-9H KIT
Catalog No. NC1704233
Encompass
X-9h Kit, 100 Capsules. Capsule kits include Dosing Applicator with Plunger, Funnel with Stand, "How to Dose" Video CD and capsules.
Catalog No. NC1704233 Supplier Torpac Inc Supplier No. X9H
X-9h Kit, 100 Capsules. Capsule kits include Dosing Applicator with Plunger, Funnel with Stand, "How to Dose" Video CD and capsules.