Learn More
Gold Biotechnology Inc X-α-Gal 1 g
SDP
Catalog No. NC2943385
Encompass_Preferred
X-α-Gal is a chromogenic substrate for α-galactosidase, yielding a blue precipitate. X-α-Gal is used for detecting two-hybrid interactions directly on the culture media. This substrate eliminates the need for β-galactosidase liquid and filter-lift assays in two-hybrid analysis.
Catalog No. NC2943385 Supplier Gold Biotechnology Inc Supplier No. XA1
X-α-Gal is a chromogenic substrate for α-galactosidase, yielding a blue precipitate. X-α-Gal is used for detecting two-hybrid interactions directly on the culture media. This substrate eliminates the need for β-galactosidase liquid and filter-lift assays in two-hybrid analysis.