Learn More
ACCU-SCOPE™ X-Cite 120 Liquid Light Guide
SDP
Catalog No. 01000954 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Type:
Liquid Light Guide
Catalog No. Type
01-000-954 Liquid Light Guide
1 options
Catalog No. 01-000-954 Supplier ACCU-SCOPE™ Supplier No. 80500038
Add to Cart
Add to Cart
Excelitas Spare Parts for Metal Halide and LED 110/120
  • Replacement liquid light guide for Excelitas X-CITE™ fluorescence illuminators
  • Compatible with X-CITE models 120 series, 120Q series, 200DC, exacte, Xylis, Xylis II, Novem, 110LED, XLED1, TURBO
Specifications
Type Liquid Light Guide
Autoclavable Not Autoclavable
Color Black
Sterility Non-sterile