Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACCU-SCOPE™ X-Cite 120 Replacement Lamp Module
SDP

Catalog No. 01000944 Shop All ACCU-SCOPE Products
Change view
Click to view available options
For Use With (Equipment):
Microscope
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-000-944 Microscope
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-000-944 Supplier ACCU-SCOPE™ Supplier No. 01263000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Excelitas Spare Parts for Metal Halide and LED 110/120. Note - Excelitas Spare Parts are non-returnable, non-cancellable.

  • Genuine Excelitas X-CITE™ 120W metal halide lamp
  • For Excelitas X-CITE 120Q series illuminators
  • 2000+ hour lifespan
  • Contains mercury

Specifications

Type Light Bulb
Color Black
For Use With (Equipment) Microscope
Voltage 120 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.