Learn More
Spectra Services Inc X-Cite exacte Replacement Lamp
SDP
Catalog No. NC1610562
Encompass
The DC powered mercury lamp used in the X‑Citeexacte and X-Cite 200DC provides convenience and excitation energy comparable to the X‑Cite 120Q lamp, with the added benefit of being more stable. Use in combination with the X-Cite exacte's Closed-Loop Feedback function for ultra-stable illumination intensity even over several days. The lamp has a guaranteed life of up to 2000* hours. This replacement lamp can be used in both the X-Cite exacte and X-Cite 200DC units only. Check your model number to be sure.
Catalog No. NC1610562 Supplier Spectra Services Inc Supplier No. 01266000R
The DC powered mercury lamp used in the X‑Citeexacte and X-Cite 200DC provides convenience and excitation energy comparable to the X‑Cite 120Q lamp, with the added benefit of being more stable. Use in combination with the X-Cite exacte's Closed-Loop Feedback function for ultra-stable illumination intensity even over several days. The lamp has a guaranteed life of up to 2000* hours. This replacement lamp can be used in both the X-Cite exacte and X-Cite 200DC units only. Check your model number to be sure.