Learn More
IBI SCIENTIFIC 3P X-Gal | 7240-90-6 | MFCD00005666 | 1 g
Supplier:  IBI SCIENTIFIC 3P IB02260
Encompass_Preferred
X-GAL - 1gm IBI X-GAL is a histochemical substrate for B-galactosidase. X-GAL is a chromogenic stain for the detection of lac gene expression.
Catalog No. NC0753460