Learn More
Medchemexpress LLC HY-15934 1g , X-GAL CAS:7240-90-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY159341G
Encompass_Preferred
HY-15934 1g X-GAL CAS: 7240-90-6 X-GAL is a widely used chromogenic β-galactosidase substrate. β-galactosidase cleaves X-gal and produce an insoluble blue compound, which is detectable. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0179