Learn More
AdipoGen X-Gal (250 mg)
SDP
Catalog No. 501687266
Encompass_Preferred
X-Gal (250 mg), Adipogen Life Sciences. Chemical. CAS: 7240-90-6. Formula: C14H15BrClNO6. MW: 408.6. Molecular biology reagent. Chromogenic substrate for beta-galactosidase. Used in gene cloning together with IPTG (Prod. No. AG-CC1-0002) to indicate whether a cell expresses the beta-galactosidase enzyme, which is encoded by the lacZ gene, in a technique called blue/white screening. Distinguishes between recombinant and non-recombinant plasmids carrying the beta-galactosidase gene. Used in two-hybrid analysis. X-Gal is used to determine E. coli and coliform content in drinking water samples.
Catalog No. 50-168-7266 Supplier AdipoGen Supplier No. AGCC10003M250
X-Gal (250 mg), Adipogen Life Sciences. Chemical. CAS: 7240-90-6. Formula: C14H15BrClNO6. MW: 408.6. Molecular biology reagent. Chromogenic substrate for beta-galactosidase. Used in gene cloning together with IPTG (Prod. No. AG-CC1-0002) to indicate whether a cell expresses the beta-galactosidase enzyme, which is encoded by the lacZ gene, in a technique called blue/white screening. Distinguishes between recombinant and non-recombinant plasmids carrying the beta-galactosidase gene. Used in two-hybrid analysis. X-Gal is used to determine E. coli and coliform content in drinking water samples.