Learn More
Alkali Scientific X a Gal [5 Bromo 4 Chloro 3 indolyl a D galactoside], 1 Gram
SDP
Catalog No. 5000660585
Encompass_Preferred
X a Gal (5-Bromo 4-Chloro 3-Indolyl α-D-Galactoside) is a chromogenic substrate used in molecular biology for the detection of β-galactosidase activity in bacterial and eukaryotic cells. This reagent is similar to X GAL but uses α-galactoside instead of β-galactoside as the substrate. When cleaved by β-galactosidase, X a Gal produces a colored product that can be easily visualized. It is commonly used in bacterial screening, gene expression assays, and enzyme activity measurements. This 1g bottle is convenient for laboratory use and allows for numerous experiments. X a Gal is particularly useful in the study of β-galactosidase function and regulation in various biological systems.
Catalog No. 50-006-60585 Supplier Alkali Scientific Supplier No. XAG001CP
X a Gal (5-Bromo 4-Chloro 3-Indolyl α-D-Galactoside) is a chromogenic substrate used in molecular biology for the detection of β-galactosidase activity in bacterial and eukaryotic cells. This reagent is similar to X GAL but uses α-galactoside instead of β-galactoside as the substrate. When cleaved by β-galactosidase, X a Gal produces a colored product that can be easily visualized. It is commonly used in bacterial screening, gene expression assays, and enzyme activity measurements. This 1g bottle is convenient for laboratory use and allows for numerous experiments. X a Gal is particularly useful in the study of β-galactosidase function and regulation in various biological systems.