Learn More
Alkali Scientific X GAL [5 Bromo 4 chloro 3 indolyl β D galactoside], 50 Grams
SDP
Catalog No. 5000661177
Encompass_Preferred
Indolyl Beta-D-galactoside (50 g) is a chromogenic substrate used in gene expression assays to detect β-galactosidase activity. This substrate undergoes hydrolysis by β-galactosidase, resulting in a blue color that can be easily detected visually or quantitatively. The 50 g formulation is ideal for large-scale research applications and is widely used in studies of gene expression, cellular metabolism, and microbial activity. It is commonly used in microbiological assays, genetic research, and diagnostic applications. X-Gal is an essential tool in molecular biology for detecting and analyzing gene activity in engineered bacteria and other organisms.
Catalog No. 50-006-61177 Supplier Alkali Scientific Supplier No. GAL008CP
Indolyl Beta-D-galactoside (50 g) is a chromogenic substrate used in gene expression assays to detect β-galactosidase activity. This substrate undergoes hydrolysis by β-galactosidase, resulting in a blue color that can be easily detected visually or quantitatively. The 50 g formulation is ideal for large-scale research applications and is widely used in studies of gene expression, cellular metabolism, and microbial activity. It is commonly used in microbiological assays, genetic research, and diagnostic applications. X-Gal is an essential tool in molecular biology for detecting and analyzing gene activity in engineered bacteria and other organisms.