Learn More
Biotang Inc X-GAL, 5 GM, P-945, CAS # 7240-90-6. Purity > 98%, soluble in DMSO or DMF. Shipped at room temperature. Store at 2-8 degrees Celsius. Mol. wt. 408.6.
Catalog No. 50152389 Shop All BIOTANG Inc Products
Encompass_Preferred
X-GAL, 5 GM, P-945, CAS # 7240-90-6. Purity > 98%, soluble in DMSO or DMF. Shipped at room temperature. Store at 2-8 degrees Celsius. Mol. wt. 408.6.
Catalog No. 50-152-389 Supplier Biotang Inc Supplier No. BTBB935
X-GAL, 5 GM, P-945, CAS # 7240-90-6. Purity > 98%, soluble in DMSO or DMF. Shipped at room temperature. Store at 2-8 degrees Celsius. Mol. wt. 408.6.