Learn More
Zymo Research Corporation 5-bromo-4-chloro-3-indolyl β-D-galactopyranoside (X-GAL) (5 ml) ^ '*'
SDP
Catalog No. 501957649
Encompass
Ready-to-use sterile 2% X-GAL solution that can be added directly to your agar plates. The X-GAL solution has been uniquely formulated to reduce false positives during blue-white screening and generates a rich blue color that is easily distinguishable from background.
Catalog No. 50-195-7649 Supplier Zymo Research Corporation Supplier No. X10015
Ready-to-use sterile 2% X-GAL solution that can be added directly to your agar plates. The X-GAL solution has been uniquely formulated to reduce false positives during blue-white screening and generates a rich blue color that is easily distinguishable from background.