Learn More
Enzo Life Sciences X-Gal (5g). CAS: 7240-90-6
SDP
Catalog No. 502009588
Encompass_Preferred
Chromogenic substrate for the enzyme β-galactosidase. Alternative name: 5-Bromo-4-chloro-3-indolyl-beta-D-galactoside. Purity: ≥98% (HPLC, TLC). Appearance: White powder. Solubility: Soluble in dimethyl formamide. Long Term Storage: -20°C.
Catalog No. 50-200-9588 Supplier Enzo Life Sciences Supplier No. ALX582002G005
Chromogenic substrate for the enzyme β-galactosidase. Alternative name: 5-Bromo-4-chloro-3-indolyl-beta-D-galactoside. Purity: ≥98% (HPLC, TLC). Appearance: White powder. Solubility: Soluble in dimethyl formamide. Long Term Storage: -20°C.