Learn More
Gold Biotechnology Inc X-Gal 10 g
SDP
Catalog No. 501532850
Encompass_Preferred
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside is most commonly known as X-Gal. X-gal is a histochemical substrate for β-galactosidase which cleaves X-Gal to yield an insoluble blue precipitate.
Catalog No. 50-153-2850 Supplier Gold Biotechnology Inc Supplier No. X4281C10
5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside is most commonly known as X-Gal. X-gal is a histochemical substrate for β-galactosidase which cleaves X-Gal to yield an insoluble blue precipitate.