Learn More
Selleck Chemical LLC X-GAL
SDP
Catalog No. 5000002843
Encompass_Preferred
X-GAL (BCIG) is a widely used chromogenic -galactosidase substrate -galactosidase cleaves X-gal and produce an insoluble detectable blue compound X-Gal is applied in blue-white selection of recombinant bacterial colonies with the lac genotype
Catalog No. 50-000-02843 Supplier Selleck Chemical LLC Supplier No. S6820100MG
X-GAL (BCIG) is a widely used chromogenic -galactosidase substrate -galactosidase cleaves X-gal and produce an insoluble detectable blue compound X-Gal is applied in blue-white selection of recombinant bacterial colonies with the lac genotype