Learn More
Research Products International Corp X-GAL 50mg/ml Solution [5-Bromo-4-Chloro-3-indoyl-B-D-galactoside solution 50mg/ml], 100 Milligrams
SDP
Catalog No. 50213185
Encompass_Preferred
Solution contains 50 mg/ml X-GAL in DMF. Inquire regarding additional concentrations.
Protect from light.
Catalog No. 50-213-185 Supplier Research Products International Corp Supplier No. B718250.1
Solution contains 50 mg/ml X-GAL in DMF. Inquire regarding additional concentrations.
Protect from light.