Learn More
Bioworld X-Gal-IPTG, Ready-to-Use Spray, 20 mL
Catalog No. 50198929
Encompass_Preferred
X-Gal-IPTG, Ready- to-use, SprayStable X-gal/IPTG non-toxic solution For use in blue/white colony screening in expression studiesDoes not contain toxic DMF and DMSOContains 40 mg/ml of IPTG (Isopropyl-β-D-Thiogalactopyranoside) and 40 mg/ml of X-gal (5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside)Supplied in a very easy to use and convenient to use Spray BottleAlso available in a Dropper Bottle (cat# 21530077-1)Use: Spray 2-3 times on desired bacterial plate Spread and ensure absorption (wait ~5-10 minutes) Plate the transformed bacteria to be tested and incubate
Catalog No. 50-198-929 Supplier Bioworld Supplier No. 215301011
X-Gal-IPTG, Ready- to-use, SprayStable X-gal/IPTG non-toxic solution For use in blue/white colony screening in expression studiesDoes not contain toxic DMF and DMSOContains 40 mg/ml of IPTG (Isopropyl-β-D-Thiogalactopyranoside) and 40 mg/ml of X-gal (5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside)Supplied in a very easy to use and convenient to use Spray BottleAlso available in a Dropper Bottle (cat# 21530077-1)Use: Spray 2-3 times on desired bacterial plate Spread and ensure absorption (wait ~5-10 minutes) Plate the transformed bacteria to be tested and incubate