Learn More
Research Products International Corp X-GAL and IPTG Ready to Use Non-Toxic Solution 10mg/ml, 500 Milligrams
SDP
Catalog No. 50488542
Encompass_Preferred
Stable solution of X-GAL and IPTG. Does not contain DMSO or DMF. Solution concentration is 10 mg/ml X-Gal and 8 mg/ml IPTG.
Chromogenic substrate of β-Galactosidase used in combination with IPTG for detection of β-Galactosidase activity in bacterial colonies. X-GAL is cleaved at the B1-4 bond between galactose and the 5-Bromo-4-chloro-3-indolyl portion of X-GAL by β-Galactosidase via hydrolysis. The cleavage of X-GAL results in the production of a water insoluble blue dichloro-dibromo-indigo precipitate. In cloning strategies with vectors such as Lambda-11, M13mp18 and 19, pUC18 and 19, pUR222 the E. coli lacZ gene is transformed to lac+ cells. WARNING! This product can expose you to 1-Methyl-2-Pyrrolidon, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-488-542 Supplier Research Products International Corp Supplier No. B715000.5
Stable solution of X-GAL and IPTG. Does not contain DMSO or DMF. Solution concentration is 10 mg/ml X-Gal and 8 mg/ml IPTG.
Chromogenic substrate of β-Galactosidase used in combination with IPTG for detection of β-Galactosidase activity in bacterial colonies. X-GAL is cleaved at the B1-4 bond between galactose and the 5-Bromo-4-chloro-3-indolyl portion of X-GAL by β-Galactosidase via hydrolysis. The cleavage of X-GAL results in the production of a water insoluble blue dichloro-dibromo-indigo precipitate. In cloning strategies with vectors such as Lambda-11, M13mp18 and 19, pUC18 and 19, pUR222 the E. coli lacZ gene is transformed to lac+ cells. WARNING! This product can expose you to 1-Methyl-2-Pyrrolidon, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.