Learn More
Bioworld X-GAL READY-TO-USE DROPPER
Supplier:  Bioworld 215301021
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. X-gal (5-Bromo-4-Chloro-3-Indolyl--D-Galactopyranoside) is a histochemical substrate widely used for beta-Galactosidase to identify lac+ bacterial colonies. It is used in conjunction with IPTG for blue/white colony screening. The solu...
Catalog No. 50-198-930