Learn More
Bioworld X-GAL READY-TO-USE SPRAY
Catalog No. 50198931
Encompass_Preferred
X-gal (5-Bromo-4-Chloro-3-Indolyl--D-Galactopyranoside) is a histochemical substrate widely used for beta-Galactosidase to identify lac+ bacterial colonies. It is used in conjunction with IPTG for blue/white colony screening. The solu...
Catalog No. 50-198-931 Supplier Bioworld Supplier No. 215301031
X-gal (5-Bromo-4-Chloro-3-Indolyl--D-Galactopyranoside) is a histochemical substrate widely used for beta-Galactosidase to identify lac+ bacterial colonies. It is used in conjunction with IPTG for blue/white colony screening. The solu...