Learn More
Research Products International Corp X-GlcA Sodium Salt [5-Bromo-4-chloro-3-indolyl-B-D-glucuronic acid, sodium salt trihydrate], 100 Milligrams
SDP
Supplier:  Research Products International Corp B721000.1
Encompass_Preferred
Histochemical substrate for β-D-Glucuronidase. Produces blue-black water insoluble color for (GUS) gene detection. Applications include use in bacterial detection systems, e.g. E. Coli.
Catalog No. 50-213-186