Learn More
Alkali Scientific X GlcA Cyclohexylammonium Salt [5 Bromo 4 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt], 100 Mg
SDP
Catalog No. 5000660465
Encompass_Preferred
X GlcA Cyclohexylammonium Salt (5 Bromo 4 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt) is a chromogenic substrate designed for laboratory use. This reagent is typically employed in assays that detect the presence of glucuronic acid derivatives in enzymatic reactions. The compound is provided in a 100 mg quantity and is widely used in biochemical research and diagnostic applications. X GlcA acts as a sensitive substrate for the detection of β-glucuronidase activity, which is important in various biological and chemical assays. This product is ideal for colorimetric detection where a visual change is used to identify enzyme activity.
Catalog No. 50-006-60465 Supplier Alkali Scientific Supplier No. XGL002CP
X GlcA Cyclohexylammonium Salt (5 Bromo 4 chloro 3 indolyl β D glucuronic acid, cyclohexylammonium salt) is a chromogenic substrate designed for laboratory use. This reagent is typically employed in assays that detect the presence of glucuronic acid derivatives in enzymatic reactions. The compound is provided in a 100 mg quantity and is widely used in biochemical research and diagnostic applications. X GlcA acts as a sensitive substrate for the detection of β-glucuronidase activity, which is important in various biological and chemical assays. This product is ideal for colorimetric detection where a visual change is used to identify enzyme activity.