Learn More
Bioworld X-Glu, 500 mg
Catalog No. 50198890
Encompass_Preferred
X-Glu (Glucopyranoside) Also known as 5-Bromo-4-Chloro-3-indolyl-β-D-Glucopyranoside. A substrate for Beta-glucosidase that renders an intense indigo-blue chromophore (615 nm) upon enzymatic action. Used as an indicatior-probe in histochemistry and in culture media to detect beta-Glucosidase positive organisms. Specifications:MF: C14H15BrClNO6MW: 408.64 CAS 15548-60-4
Catalog No. 50-198-890 Supplier Bioworld Supplier No. 215300792
X-Glu (Glucopyranoside) Also known as 5-Bromo-4-Chloro-3-indolyl-β-D-Glucopyranoside. A substrate for Beta-glucosidase that renders an intense indigo-blue chromophore (615 nm) upon enzymatic action. Used as an indicatior-probe in histochemistry and in culture media to detect beta-Glucosidase positive organisms. Specifications:MF: C14H15BrClNO6MW: 408.64 CAS 15548-60-4