Learn More
Carbosynth LLC. 5-Bromo-4-chloro-3-indoxyl-beta-D-glucuronic acid, cyclohexylammonium salt monohydrate, 114162-64-0, MFCD00058543, 1 g
SDP
Catalog No. 50851897
Encompass_Preferred
Formula: C20H28BrClN2O8, Molecular weight: 539.81 g/mol, 99%, Store at -15°C, Synonyms: X-beta-D-glucuronide cyclohexylammonium salt; X-beta-D-glucuronide CHX-salt; X-beta-D-GlcA · CHX.
Catalog No. 50-851-897 Supplier Carbosynth LLC. Supplier No. B73001G
Formula: C20H28BrClN2O8, Molecular weight: 539.81 g/mol, 99%, Store at -15°C, Synonyms: X-beta-D-glucuronide cyclohexylammonium salt; X-beta-D-glucuronide CHX-salt; X-beta-D-GlcA · CHX.