Learn More
Gold Biotechnology Inc X-gluc (CHX salt) 100 mg
SDP
Catalog No. 501532774
Encompass_Preferred
X-Gluc is a chromogenic substrate for beta-glucuronidase (GUS) which is encoded by the gusA (uid A) gene, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate. X-Gluc is often used in GUS staining of plant cells and tissue and in infections caused by E. coli.
Catalog No. 50-153-2774 Supplier Gold Biotechnology Inc Supplier No. G1281C
X-Gluc is a chromogenic substrate for beta-glucuronidase (GUS) which is encoded by the gusA (uid A) gene, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate. X-Gluc is often used in GUS staining of plant cells and tissue and in infections caused by E. coli.