Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC X-gluc (cyclohexanamine) | 114162-64-0 | MFCD00058543 | 99.8% | 521.79 g·mol⁻1 | C20H26BrClN2O7 | 10 MG
SDP

Catalog No. 5000288108
Encompass_Preferred

X-gluc (cyclohexanamine) is a chromogenic substrate for β-glucuronidase (GUS). When enzymatically cleaved it produces an insoluble blue precipitate, enabling visual detection of GUS activity in microbiological and molecular biology assays.

  • Chromogenic substrate that yields a blue precipitate upon GUS cleavage.
  • High purity (≈99.8%), suitable for research applications.
  • Solid, white to off-white appearance for easy handling.
  • Soluble in DMSO up to 100 mg/mL; ultrasonic assistance may be required.
  • Recommended storage: 4°C, protected from light; in solution store at -80°C for long term.
  • Supplied in small pack sizes for assay-scale experiments.
  • Intended for research use only.

Catalog No. 50-002-88108 Supplier Medchemexpress LLC Supplier No. HY15935B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

X-gluc (cyclohexanamine) is a chromogenic substrate for β-glucuronidase (GUS). When enzymatically cleaved it produces an insoluble blue precipitate, enabling visual detection of GUS activity in microbiological and molecular biology assays.

  • Chromogenic substrate that yields a blue precipitate upon GUS cleavage.
  • High purity (≈99.8%), suitable for research applications.
  • Solid, white to off-white appearance for easy handling.
  • Soluble in DMSO up to 100 mg/mL; ultrasonic assistance may be required.
  • Recommended storage: 4°C, protected from light; in solution store at -80°C for long term.
  • Supplied in small pack sizes for assay-scale experiments.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.