Learn More
Bioworld X-Gluc, 1 g
Catalog No. 50198894
Encompass_Preferred
X-glucuronide, Cyclohexylammonium Salt Also known as 5-Bromo-4-Chloro-3-indolyl-β-D-Glucuronic acid, Cyclohexylammonium salt. A widely used histochemical substrate for β-glucoronidase (GUS). Used for the detection of GUS gene in bacteria or for histochemical applications in plant tissues as it is converted by the enzyme into an intense blue precipitate.MF: C14H13BrClNO7*C6H13NCAS: 114162-64-0CAS: 18656-96-7Ultra pure gradeAllow to warm to room temperature before opening.Soluble in DMSO and DMFO
Catalog No. 50-198-894 Supplier Bioworld Supplier No. 215300804
X-glucuronide, Cyclohexylammonium Salt Also known as 5-Bromo-4-Chloro-3-indolyl-β-D-Glucuronic acid, Cyclohexylammonium salt. A widely used histochemical substrate for β-glucoronidase (GUS). Used for the detection of GUS gene in bacteria or for histochemical applications in plant tissues as it is converted by the enzyme into an intense blue precipitate.MF: C14H13BrClNO7*C6H13NCAS: 114162-64-0CAS: 18656-96-7Ultra pure gradeAllow to warm to room temperature before opening.Soluble in DMSO and DMFO