Learn More
Bioworld X-Gluc Enhancer Solution, 50 mL
Catalog No. 50198859
Encompass_Preferred
Staining solution containing:X-Gluc (5-bromo-4-chloro-3-indolyl-β-D-glucuronide 100 mM phosphate buffer, pH 7.0 5 mM K3[Fe(CN)6]5 mM K4[Fe(CN)6]10 mM EDTA
Catalog No. 50-198-859 Supplier Bioworld Supplier No. 215300542
Staining solution containing:X-Gluc (5-bromo-4-chloro-3-indolyl-β-D-glucuronide 100 mM phosphate buffer, pH 7.0 5 mM K3[Fe(CN)6]5 mM K4[Fe(CN)6]10 mM EDTA