Learn More
MSC Zep Floor Degreaser, Alkaline, Heavy Duty, Low Odor, 55 gal
Catalog No. 502867378
Encompass
Zep Extra is a fast-acting, heavy-duty industrial floor cleaner and degreaser designed to remove dirt, grease, and heavy soil from concrete, ceramic tile, terrazzo, and masonry floors. Its high-performance, low-foam formula works effectively in manual cleaning and auto-scrubbing machines, and is approved for use in federally inspected meat and poultry processing plants.
  • Effectively removes dirt, grease, heavy soil, carbon, soot, and ink
  • Suitable for concrete, masonry, brick, and ceramic tile floors
  • Compatible with manual cleaning methods and auto-scrubbing machines
  • Approved for use in federally inspected meat and poultry processing plants
  • Customizable dilution based on soil severity for versatile applications
  • Formulated for industrial and business use only
Catalog No. 50-286-7378 Supplier MSC Supplier No. 20501078
Zep Extra is a fast-acting, heavy-duty industrial floor cleaner and degreaser designed to remove dirt, grease, and heavy soil from concrete, ceramic tile, terrazzo, and masonry floors. Its high-performance, low-foam formula works effectively in manual cleaning and auto-scrubbing machines, and is approved for use in federally inspected meat and poultry processing plants.
  • Effectively removes dirt, grease, heavy soil, carbon, soot, and ink
  • Suitable for concrete, masonry, brick, and ceramic tile floors
  • Compatible with manual cleaning methods and auto-scrubbing machines
  • Approved for use in federally inspected meat and poultry processing plants
  • Customizable dilution based on soil severity for versatile applications
  • Formulated for industrial and business use only