Learn More
MOLECULAR DEPOT LLC X-Neu5Ac - 1 mg
Catalog No. 5000013591
Encompass_Preferred
X-Neu5Ac (X-Sialic acid X-NANA) is a synthetic 5-Bromo-4-Chloroindol-3-yl-5-Acetamido-3 5-Dideoxy-alpha-D-Glycero-D-Galacto-2-Nonulopyranosidonic Acid The molecular weight of this peptide is 537 74 g/mol This product is for in vitro research use only This product is not for human or animal use
Catalog No. 50-000-13591 Supplier MOLECULAR DEPOT LLC Supplier No. MDP0878
X-Neu5Ac (X-Sialic acid X-NANA) is a synthetic 5-Bromo-4-Chloroindol-3-yl-5-Acetamido-3 5-Dideoxy-alpha-D-Glycero-D-Galacto-2-Nonulopyranosidonic Acid The molecular weight of this peptide is 537 74 g/mol This product is for in vitro research use only This product is not for human or animal use