Learn More
TARGETMOL CHEMICALS INC X-NEUNAC 25MG
SDP
Catalog No. 502577217
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. X-NeuNAc (5-Bromo-4-chloro-3-indolyl-alpha-D-N-acetylneuraminic acid sodium salt) is a new chromogenic assay substrate of neuraminidase activity in bacterial expression systems. purity: 98%
Catalog No. 50-257-7217 Supplier TARGETMOL CHEMICALS INC Supplier No. T2148625MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. X-NeuNAc (5-Bromo-4-chloro-3-indolyl-alpha-D-N-acetylneuraminic acid sodium salt) is a new chromogenic assay substrate of neuraminidase activity in bacterial expression systems. purity: 98%