Learn More
Research Products International Corp X-Phos, BCIP, 5 G
SDP
Catalog No. 501976873
Encompass_Preferred
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques. Forms a purple insoluble precipitate when used in conjunction with NBT.
Catalog No. 50-197-6873 Supplier Research Products International Corp Supplier No. B740005.0
Colorimetric substrate for Alkaline phosphatase activity in blotting immunohistochemical and cytochemistry techniques. Forms a purple insoluble precipitate when used in conjunction with NBT.