Learn More
USA Scientific Inc X-Pierce cross-cut pierceable film, sterile
Catalog No. NC9579640
Encompass
X-Pierce hairline cross-cut pierceable film with memory flaps, -40 to +90C, sterile, 50/box
Catalog No. NC9579640 Supplier USA Scientific Inc Supplier No. 29970110
X-Pierce hairline cross-cut pierceable film with memory flaps, -40 to +90C, sterile, 50/box