Learn More
Research Products International Corp X-Pierce Film, Sterile, 50 per Package
SDP
Catalog No. 50998794
Encompass
Film allows easy penetration of probe/tip without adhesive interference. Ideally designed film features pre-cut 'X' pattern for 96 well plates. 'X' flaps prevent well contents from cross contamination during multiple pipetting. Special flaps quickly return to original position after sampling. Film is 100um thick vinyl with 20um adhesive. X-Pierce is a Trademark of Excel Scientific, Inc.
Catalog No. 50-998-794 Supplier Research Products International Corp Supplier No. 202541
Film allows easy penetration of probe/tip without adhesive interference. Ideally designed film features pre-cut 'X' pattern for 96 well plates. 'X' flaps prevent well contents from cross contamination during multiple pipetting. Special flaps quickly return to original position after sampling. Film is 100um thick vinyl with 20um adhesive. X-Pierce is a Trademark of Excel Scientific, Inc.