Learn More
Andwin Scientific X-PIERCE FILMS NONSTERILE
SDP
Catalog No. NC9920880
Encompass_Preferred
The X-Pierce film is constructed with 100 m thick vinyl. It is ideal for protecting contents of plate from contamination during multiple manual reagent additions over time. A precut "X" over each well conveniently produces four flaps that easily bend downward when pushed by a robotic probe or pipette tip, permitting sample access without coring or adhesive fouling. The flaps return to their original position after sampling to provide continued protection. This product is nonsterile.
Catalog No. NC9920880 Supplier Andwin Scientific Supplier No. XP100
The X-Pierce film is constructed with 100 m thick vinyl. It is ideal for protecting contents of plate from contamination during multiple manual reagent additions over time. A precut "X" over each well conveniently produces four flaps that easily bend downward when pushed by a robotic probe or pipette tip, permitting sample access without coring or adhesive fouling. The flaps return to their original position after sampling to provide continued protection. This product is nonsterile.